siRNA Details
| Gene Symbol: ZNF137P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023311.1 | 3 | 2333 | |||
| GenBank mRNA | U09414.1 | - | 3 | 2367 | ||
| Gene Symbol: ZNF816-ZNF321P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR749224.1 | - | 5 | 1957 | ||

ZNF137P
Genomic Map