siRNA Details
Gene Symbol: ZNF137P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023311.1 | 2 | 994 | |||
GenBank mRNA | BC110455.1 | - | 2 | 994 | ||
BC110456.1 | - | 2 | 995 | |||
BQ893730.1 | - | 1 | 159 | |||
U09414.1 | - | 2 | 1027 |
