siRNA Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054818.1 | - | 1 | 1515 | ||
| AK125445.1 | - | 1 | 2362 | |||
| AK128507.1 | - | 1 | 3350 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG765895.1 | - | 1 | 802 | Chr. 1 - 2190119 - 2191031 [-] on Build GRCh38 | |

FAAP20
Genomic Map