siRNA Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256946.1 | NP_001243875.1 | 4 | 978 | ||
| GenBank mRNA | AK125445.1 | - | 1 | 101 | ||
| AK128507.1 | - | 1 | 1091 | |||
| BC041041.1 | - | 4 | 917 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAAP20| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256946.1 | NP_001243875.1 | 4 | 978 | ||
| GenBank mRNA | AK125445.1 | - | 1 | 101 | ||
| AK128507.1 | - | 1 | 1091 | |||
| BC041041.1 | - | 4 | 917 | |||