siRNA Details
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054818.1 | - | 1 | 584 | ||
AK125445.1 | - | 1 | 1431 | |||
AK128507.1 | - | 1 | 2419 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054818.1 | - | 1 | 584 | ||
AK125445.1 | - | 1 | 1431 | |||
AK128507.1 | - | 1 | 2419 |