siRNA Details
| Gene Symbol: PCAT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024259.1 | 2 | 1863 | |||
| GenBank mRNA | AK056805.1 | - | 2 | 1863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCAT18| Gene Symbol: PCAT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024259.1 | 2 | 1863 | |||
| GenBank mRNA | AK056805.1 | - | 2 | 1863 | ||