siRNA Details
| Gene Symbol: PCAT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024259.1 | 2 | 453 | |||
| GenBank mRNA | AK056805.1 | - | 2 | 453 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA865211.1 | - | 2 | 453 | Chr. 18 - 26689394 - 26703638 [-] on Build GRCh38 | |

PCAT18
Genomic Map