siRNA Details
| Gene Symbol: LOC400927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002821.2 | 5 | 480 | |||
| GenBank mRNA | CN357790.1 | - | 5 | 392 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA847599.1 | - | 5 | 427 | Chr. 22 - 38344532 - 38398873 [-] on Build GRCh38 | |

LOC400927
Genomic Map