siRNA Details
| Gene Symbol: LOC728613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003713.1 | 4 | 2559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728613| Gene Symbol: LOC728613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003713.1 | 4 | 2559 | |||