siRNA Details
| Gene Symbol: LOC728613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003713.1 | 4 | 1000 | |||
| GenBank mRNA | AK311210.1 | - | 4 | 998 | ||
| BC014604.1 | - | 5 | 825 | |||
| BC045555.1 | - | 4 | 989 | |||
| BC053586.1 | - | 7 | 2056 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728613| Gene Symbol: LOC728613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003713.1 | 4 | 1000 | |||
| GenBank mRNA | AK311210.1 | - | 4 | 998 | ||
| BC014604.1 | - | 5 | 825 | |||
| BC045555.1 | - | 4 | 989 | |||
| BC053586.1 | - | 7 | 2056 | |||