siRNA Details
| Gene Symbol: LOC646762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024278.1 | 2 | 3524 | |||
| GenBank mRNA | AK092870.1 | - | 2 | 1414 | ||
| BC044638.1 | - | 1 | 1824 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC646762| Gene Symbol: LOC646762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024278.1 | 2 | 3524 | |||
| GenBank mRNA | AK092870.1 | - | 2 | 1414 | ||
| BC044638.1 | - | 1 | 1824 | |||