siRNA Details
| Gene Symbol: ITPK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002808.2 | 1 | 524 | |||
| GenBank mRNA | AK024887.1 | - | 1 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPK1-AS1| Gene Symbol: ITPK1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002808.2 | 1 | 524 | |||
| GenBank mRNA | AK024887.1 | - | 1 | 526 | ||