siRNA Details
| Gene Symbol: LOC100130417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026874.2 | 3 | 532 | |||
| NR_122045.1 | 3 | 472 | ||||
| GenBank mRNA | AK096928.1 | - | 1 | 2924 | ||
| BC025320.1 | - | 1 | 578 | |||
| BC144023.1 | - | 3 | 277 | |||
| BC144024.1 | - | 3 | 217 | |||

LOC100130417
Genomic Map