siRNA Details
| Gene Symbol: LOC100130417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026874.2 | 1 | 266 | |||
| NR_122045.1 | 1 | 266 | ||||
| GenBank mRNA | AK096928.1 | - | 1 | 1590 | ||
| BC144023.1 | - | 11 | ||||
| BC144024.1 | - | 11 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130417| Gene Symbol: LOC100130417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026874.2 | 1 | 266 | |||
| NR_122045.1 | 1 | 266 | ||||
| GenBank mRNA | AK096928.1 | - | 1 | 1590 | ||
| BC144023.1 | - | 11 | ||||
| BC144024.1 | - | 11 | ||||