siRNA Details
| Gene Symbol: NOC2LP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002826.2 | 1 | 106 | |||
| GenBank mRNA | AK302229.1 | - | 1 | 106 | ||
| BC060851.1 | - | 1 | 95 | |||
| Gene Symbol: NOC2LP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK297320.1 | - | 1 | 80 | ||

NOC2LP2
Genomic Map