siRNA Details
| Gene Symbol: NOC2LP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002826.2 | 1 | 653 | |||
| GenBank mRNA | AK302229.1 | - | 1 | 653 | ||
| BC060851.1 | - | 1 | 646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOC2LP2| Gene Symbol: NOC2LP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002826.2 | 1 | 653 | |||
| GenBank mRNA | AK302229.1 | - | 1 | 653 | ||
| BC060851.1 | - | 1 | 646 | |||