siRNA Details
| Gene Symbol: FAM225A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024366.1 | 3 | 5461 | |||
| GenBank mRNA | AK127516.1 | - | 1 | 4447 | ||
| AL832752.1 | - | 3 | 5284 | |||
| BX647840.1 | - | 2 | 5688 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG164717.1 | - | 1 | 618 | Chr. 9 - 113118888 - 113119677 [+] on Build GRCh38 | |

FAM225A
Genomic Map