siRNA Details
| Gene Symbol: C14orf132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252507.2 | NP_001239436.1 | 2 | 2831 | ||
| NM_001282463.1 | NP_001269392.1 | 3 | 2930 | |||
| NM_001289139.1 | NP_001276068.1 | 3 | 2924 | |||
| XM_017021469.1 | XP_016876958.1 | 2 | 4163 | |||
| GenBank mRNA | AK001590.1 | - | 2 | 2774 | ||
| AK025915.1 | - | 1 | 635 | |||
| AK074462.1 | - | 1 | 1769 | |||
| BC043593.1 | - | 2 | 2814 | |||

C14orf132
Genomic Map