siRNA Details
| Gene Symbol: C14orf132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252507.2 | NP_001239436.1 | 2 | 5023 | ||
| NM_001282463.1 | NP_001269392.1 | 3 | 5122 | |||
| NM_001289139.1 | NP_001276068.1 | 3 | 5116 | |||
| XM_017021469.1 | XP_016876958.1 | 2 | 6355 | |||
| GenBank mRNA | AL390130.1 | - | 1 | 901 | ||
| BC042922.1 | - | 1 | 902 | |||
| BC043593.1 | - | 2 | 5004 | |||

C14orf132
Genomic Map