siRNA Details
| Gene Symbol: CRHR1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026905.2 | 1 | 181 | |||
| NR_026906.2 | 1 | 181 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG534719.1 | - | 1 | 182 | Chr. 17 - 45638975 - 45646008 [+] on Build GRCh38 | |

CRHR1-IT1
Genomic Map