siRNA Details
| Gene Symbol: PP14571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024014.1 | 2 | 1945 | |||
| GenBank mRNA | AF070536.1 | - | 1 | 1356 | ||
| AK092442.1 | - | 2 | 1945 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PP14571| Gene Symbol: PP14571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024014.1 | 2 | 1945 | |||
| GenBank mRNA | AF070536.1 | - | 1 | 1356 | ||
| AK092442.1 | - | 2 | 1945 | |||