siRNA Details
| Gene Symbol: ZBED3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024398.1 | 2 | 135 | |||
| GenBank mRNA | AK311179.1 | - | 2 | 166 | ||
| CX786597.1 | - | 2 | 192 | |||
| DA293448.1 | - | 2 | 138 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBED3-AS1| Gene Symbol: ZBED3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024398.1 | 2 | 135 | |||
| GenBank mRNA | AK311179.1 | - | 2 | 166 | ||
| CX786597.1 | - | 2 | 192 | |||
| DA293448.1 | - | 2 | 138 | |||