siRNA Details
Gene Symbol: ZBED3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024398.1 | 4 | 1124 | |||
GenBank mRNA | AK055144.1 | - | 5 | 1547 | ||
AK098256.1 | - | 3 | 1794 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZBED3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024398.1 | 4 | 1124 | |||
GenBank mRNA | AK055144.1 | - | 5 | 1547 | ||
AK098256.1 | - | 3 | 1794 |