siRNA Details
| Gene Symbol: LOC286359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026847.1 | 6 | 1382 | |||
| GenBank mRNA | AK097847.1 | - | 1 | 1794 | ||
| AL833510.1 | - | 4 | 2823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC286359| Gene Symbol: LOC286359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026847.1 | 6 | 1382 | |||
| GenBank mRNA | AK097847.1 | - | 1 | 1794 | ||
| AL833510.1 | - | 4 | 2823 | |||