siRNA Details
| Gene Symbol: LOC286359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026847.1 | 5 | 561 | |||
| GenBank mRNA | AK097847.1 | - | 1 | 597 | ||
| AK310275.1 | - | 3 | 344 | |||
| AK310296.1 | - | 5 | 561 | |||
| AL833510.1 | - | 4 | 1626 | |||
| BC105125.1 | - | 1 | 509 | |||
| BC112056.1 | - | 1 | 509 | |||

LOC286359
Genomic Map