siRNA Details
| Gene Symbol: LOC646813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB231713.1 | - | 5 | 603 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC646813| Gene Symbol: LOC646813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB231713.1 | - | 5 | 603 | ||