siRNA Details
| Gene Symbol: NIPSNAP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018376.3 | NP_060846.2 | 6 | 3092 | ||
| NR_130759.1 | 5 | 2881 | ||||
| XM_011518839.1 | XP_011517141.1 | 5 | 2942 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NIPSNAP3B| Gene Symbol: NIPSNAP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018376.3 | NP_060846.2 | 6 | 3092 | ||
| NR_130759.1 | 5 | 2881 | ||||
| XM_011518839.1 | XP_011517141.1 | 5 | 2942 | |||