siRNA Details
| Gene Symbol: FKBP9P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003949.1 | 6 | 947 | |||
| NR_027339.1 | 4 | 1051 | ||||
| NR_027340.1 | 3 | 680 | ||||
| NR_027342.1 | 4 | 961 | ||||
| GenBank mRNA | BC011872.2 | - | 4 | 961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FKBP9P1| Gene Symbol: FKBP9P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003949.1 | 6 | 947 | |||
| NR_027339.1 | 4 | 1051 | ||||
| NR_027340.1 | 3 | 680 | ||||
| NR_027342.1 | 4 | 961 | ||||
| GenBank mRNA | BC011872.2 | - | 4 | 961 | ||