siRNA Details
| Gene Symbol: LINC00632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028344.1 | 2 | 72 | |||
| NR_028345.2 | 2 | 72 | ||||
| NR_104228.1 | 2 | 72 | ||||
| GenBank mRNA | AK054978.1 | - | 2 | 64 | ||
| BM811380.1 | - | 2 | 51 | |||
| BX508035.1 | - | 2 | 65 | |||
| DA082671.1 | - | 2 | 72 | |||
| DA128399.1 | - | 2 | 72 | |||

LINC00632
Genomic Map