siRNA Details
| Gene Symbol: FLJ42393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024413.1 | 1 | 1364 | |||
| GenBank mRNA | AK124384.1 | - | 1 | 1362 | ||
| BC132980.1 | - | 1 | 752 | |||
| BC132982.1 | - | 1 | 752 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ42393| Gene Symbol: FLJ42393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024413.1 | 1 | 1364 | |||
| GenBank mRNA | AK124384.1 | - | 1 | 1362 | ||
| BC132980.1 | - | 1 | 752 | |||
| BC132982.1 | - | 1 | 752 | |||