siRNA Details
| Gene Symbol: LOC107133515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015341.2 | 9 | 4232 | |||
| Gene Symbol: LRRC37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123784.1 | - | 1 | 2094 | ||
| Gene Symbol: LRRC37BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL713754.1 | - | 2 | 3495 | ||
| BC035414.1 | - | 1 | 848 | |||

LOC107133515
Genomic Map