siRNA Details
| Gene Symbol: SBDSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001588.2 | 1 | 103 | |||
| NR_024109.1 | 1 | 103 | ||||
| NR_024110.1 | 1 | 103 | ||||
| NR_024111.1 | 1 | 103 | ||||
| GenBank mRNA | DA143511.1 | - | 1 | 103 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BP257376.1 | - | 1 | 89 | Chr. 7 - 72829388 - 72831664 [+] on Build GRCh38 | |

SBDSP1
Genomic Map