siRNA Details
Gene Symbol: RNF217-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026876.1 | 2 | 218 | |||
GenBank mRNA | AL713649.2 | - | 2 | 218 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF217-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026876.1 | 2 | 218 | |||
GenBank mRNA | AL713649.2 | - | 2 | 218 |