siRNA Details
| Gene Symbol: OR6W1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002140.1 | 1 | 654 | |||
| GenBank mRNA | BC069382.1 | - | 1 | 245 | ||
| BC074961.2 | - | 1 | 230 | |||
| BC074962.2 | - | 1 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6W1P| Gene Symbol: OR6W1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002140.1 | 1 | 654 | |||
| GenBank mRNA | BC069382.1 | - | 1 | 245 | ||
| BC074961.2 | - | 1 | 230 | |||
| BC074962.2 | - | 1 | 230 | |||