siRNA Details
| Gene Symbol: LINC00597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026813.1 | 1 | 220 | |||
| GenBank mRNA | AF216224.1 | - | 1 | 226 | ||
| BC069765.1 | - | 1 | 156 | |||
| BC103988.1 | - | 1 | 146 | |||
| BC103989.1 | - | 1 | 146 | |||
| BC103990.1 | - | 1 | 146 | |||
| BC103991.1 | - | 1 | 146 | |||

LINC00597
Genomic Map