siRNA Details
| Gene Symbol: LINC00623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024584.2 | 1 | 56 | |||
| GenBank mRNA | BC089397.1 | - | 1 | 55 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00623| Gene Symbol: LINC00623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024584.2 | 1 | 56 | |||
| GenBank mRNA | BC089397.1 | - | 1 | 55 | ||