siRNA Details
| Gene Symbol: LINC00882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028303.1 | 2 | 223 | |||
| GenBank mRNA | BX117271.1 | - | 2 | 141 | ||
| DB443285.1 | - | 2 | 223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00882| Gene Symbol: LINC00882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028303.1 | 2 | 223 | |||
| GenBank mRNA | BX117271.1 | - | 2 | 141 | ||
| DB443285.1 | - | 2 | 223 | |||