siRNA Details
| Gene Symbol: BRE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028308.1 | 1 | 1085 | |||
| GenBank mRNA | BC014088.2 | - | 1 | 1059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRE-AS1| Gene Symbol: BRE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028308.1 | 1 | 1085 | |||
| GenBank mRNA | BC014088.2 | - | 1 | 1059 | ||