siRNA Details
| Gene Symbol: BRE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028308.1 | 1 | 1620 | |||
| GenBank mRNA | DR980668.1 | - | 1 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRE-AS1| Gene Symbol: BRE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028308.1 | 1 | 1620 | |||
| GenBank mRNA | DR980668.1 | - | 1 | 241 | ||