siRNA Details
| Gene Symbol: LINC00598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024505.1 | 3 | 217 | |||
| NR_024506.1 | 3 | 217 | ||||
| NR_024507.2 | 3 | 217 | ||||
| GenBank mRNA | AK025090.1 | - | 1 | 2802 | ||
| AY116214.1 | - | 3 | 217 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00598| Gene Symbol: LINC00598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024505.1 | 3 | 217 | |||
| NR_024506.1 | 3 | 217 | ||||
| NR_024507.2 | 3 | 217 | ||||
| GenBank mRNA | AK025090.1 | - | 1 | 2802 | ||
| AY116214.1 | - | 3 | 217 | |||