siRNA Details
| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026856.1 | 3 | 1269 | |||
| GenBank mRNA | AK055096.1 | - | 1 | 2166 | ||
| AK123663.1 | - | 2 | 2504 | |||
| BC108683.1 | - | 3 | 1484 | |||
| BX640709.1 | - | 3 | 1267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01011| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026856.1 | 3 | 1269 | |||
| GenBank mRNA | AK055096.1 | - | 1 | 2166 | ||
| AK123663.1 | - | 2 | 2504 | |||
| BC108683.1 | - | 3 | 1484 | |||
| BX640709.1 | - | 3 | 1267 | |||