siRNA Details
| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026856.1 | 3 | 597 | |||
| GenBank mRNA | AK055096.1 | - | 1 | 1494 | ||
| AK123663.1 | - | 2 | 1832 | |||
| BC108683.1 | - | 3 | 812 | |||
| BC137372.1 | - | 2 | 1289 | |||
| BX640709.1 | - | 3 | 595 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI820564.1 | - | 3 | 609 | Chr. 6 - 2987981 - 2990348 [+] on Build GRCh38 | |

LINC01011
Genomic Map