siRNA Details
| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026856.1 | 3 | 649 | |||
| GenBank mRNA | AK055096.1 | - | 1 | 1546 | ||
| AK123663.1 | - | 2 | 1884 | |||
| BC108683.1 | - | 3 | 864 | |||
| BC137372.1 | - | 2 | 1341 | |||
| BX640709.1 | - | 3 | 647 | |||

LINC01011
Genomic Map