siRNA Details
| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026856.1 | 3 | 735 | |||
| GenBank mRNA | AK055096.1 | - | 1 | 1632 | ||
| AK123663.1 | - | 2 | 1970 | |||
| BC108683.1 | - | 3 | 950 | |||
| BC137372.1 | - | 2 | 1427 | |||
| BX640709.1 | - | 3 | 733 | |||

LINC01011
Genomic Map