siRNA Details
| Gene Symbol: LINC01011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026856.1 | 2 | 120 | |||
| GenBank mRNA | AK055096.1 | - | 1 | 884 | ||
| AK123663.1 | - | 2 | 1220 | |||
| BC108683.1 | - | 2 | 309 | |||
| BC137372.1 | - | 1 | 786 | |||
| BX640709.1 | - | 2 | 118 | |||
| DA076107.1 | - | 2 | 120 | |||

LINC01011
Genomic Map