siRNA Details
| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282446.1 | NP_001269375.1 | 3 | 2724 | ||
| GenBank mRNA | AK096857.1 | - | 1 | 2560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

YAE1D1| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282446.1 | NP_001269375.1 | 3 | 2724 | ||
| GenBank mRNA | AK096857.1 | - | 1 | 2560 | ||