siRNA Details
Gene Symbol: LINC00336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027908.1 | 2 | 629 | |||
GenBank mRNA | AK125740.1 | - | 2 | 629 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027908.1 | 2 | 629 | |||
GenBank mRNA | AK125740.1 | - | 2 | 629 |