siRNA Details
| Gene Symbol: LINC00242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026781.1 | 2 | 502 | |||
| GenBank mRNA | AK056013.1 | - | 2 | 502 | ||
| BC119778.1 | - | 1 | 185 | |||
| BC122871.1 | - | 1 | 186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00242| Gene Symbol: LINC00242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026781.1 | 2 | 502 | |||
| GenBank mRNA | AK056013.1 | - | 2 | 502 | ||
| BC119778.1 | - | 1 | 185 | |||
| BC122871.1 | - | 1 | 186 | |||