siRNA Details
| Gene Symbol: ZNF815P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023382.1 | 7 | 2700 | |||
| GenBank mRNA | AK096288.1 | - | 7 | 2699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF815P| Gene Symbol: ZNF815P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023382.1 | 7 | 2700 | |||
| GenBank mRNA | AK096288.1 | - | 7 | 2699 | ||