siRNA Details
| Gene Symbol: SNORD15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_000025.1 | 1 | 108 | |||
| GenBank mRNA | L15557.1 | - | 1 | 108 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA636472.1 | - | 104 | Chr. - - [-] on Build | ||

SNORD15B
Genomic Map